Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TRPM7 Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
This antibody does not react with overexpressed mouse TRPM7, and does not work on endonous samples tested for Western analysis (human, rat and mouse cell lines). The TRPM7 (Transient Receptor Protein-PhosphoLipase C-Interacting Kinase) protein is a member of the ion channel proteins. It regulates the calcium concentration in cells, which is imperative for such functions as muscle contractions and neuronal firing. TRPM7, however, is unique among the TRP channel proteins because it is bi-functional. It not only controls ion channels but it also acts as a kinase, phosphorylating proteins.

Type: Primary
Antigen: TRPM7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µL 100 µl PIEAPA116856 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol TRPM7
Antigen name Transient Receptor Potential Cation Channel Subfamily M member 7
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoPrecipitation Yes
Antigen synonyms tr, FLJ20117, LTRPC7, CHAK, LTRPC ion channel family member 7, CHAK1, homolog of mouse transient receptor potential-phospholipase C-interacting kinase
Gene ID 54822
Format whole serum
Immunogen A synthetic peptide made to the C-terminal region of rat TRPM7.
Form Liquid
Storage buffer whole serum, 0.05% sodium azide
Storage temperature -20° C, Avoid Freeze/Thaw Cycles
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty