Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-WNK1 Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
In Western blot, this antibody recognizes the short form of WNK1, ~190 kDa. The theoretical molecular weight of human WNK1 is ~251 kDa and rat WNK1 is ~230 kDa. WNK kinases play an important role in hypertension. WNK1 is a cytoplasmic protein that localizes to a segment of kidney involved in salt, potassium, and pH homeostasis, the distal nephron.

Type: Primary
Antigen: WNK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Mouse




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µL 100 µl PIEAPA116871 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol WNK1
Antigen name WNK lysine deficient Protein kinase 1
Reactivity Rat, Human, Mouse
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoFluorescence Yes
Antigen synonyms Protein kinase with no lysine 1, Kinase deficient protein, Serine/threonine protein kinase WNK1, Protein kinase lysine deficient 1
Gene ID 232341, 116477, 65125
Concentration 5 mg/ml
Immunogen A synthetic peptide sequence within the C-teminal region of human WNK1 (within the last 350 amino acids).
Purification Antigen affinity chromatography
Form Liquid
Storage buffer PBS, 0.05% sodium azide
Storage temperature -20° C, Avoid Freeze/Thaw Cycles
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty