Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-beta Actin Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
This antibody will not work in ICC/IF applications. Suggested positive control: Hela whole cell extract. In vertebrates 3 main groups of actin isoforms, alpha, beta and gamma, have been identified. The alpha actins are found in muscle tissues and are a major constituent of contractile apparatus. The beta and gamma actins coexist in most cell types as components of the cyto-skeleton and as mediators of internal cell motility. Beta actins are hightly conserved proteins that are involved in cell motility, structure and integrity. Beta actins are cytoplasmic proteins ubuquitously expressed in all eukaryotic cells. Polymerization of globular actin (G-actin) leads to a structural filament (F-actin) in the form of two stranded helix. Each actin can bind to 4 others. This antibody serves as an excellent loading control.

Type: Primary
Antigen: beta Actin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µg 100 µl PIEAPA116889 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol beta Actin
Antigen name Actin beta
Reactivity Mouse, Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
Antigen synonyms beta actin, gamma actin, Atcb, cytoskeleton actin
Gene ID 60, 11461
Concentration 1 mg/ml
Immunogen N-terminus of human beta actin using the numbering given in Swiss-prot entry P60709 and is highly conserved with gamma actin
Purification Antigen affinity chromatography
Form Liquid
Storage buffer tris citrate, tris phosphate, 0.09% sodium azide, pH 7-8
Storage temperature 4° C, do not freeze
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping