Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LRP8 Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
PA1-16913 is specific for the ~106 kDa (membrane form) ApoER2 protein. A larger ~130 kDa band may also be seen (glycosylated form). Suggested positive control: mouse brain membrane preparation. Apolipoprotein E receptor 2 (ApoER2) is an NMDA associated protein found in neurons. It participates in important functions from guiding neurons in the embryonic brain to regulating synaptic signaling. It is also involved in debilitating neurodegenerative diseases such as schizophrenia and Alzheimer's disease.

Type: Primary
Antigen: LRP8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Human, Chicken




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µL 100 µl PIEAPA116913 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol LRP8
Antigen name Apolipoprotein E Receptor 2
Reactivity Chicken, Mouse, Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoFluorescence Yes
Antigen synonyms Low-density lipoprotein receptor-related protein 8 antibody, Apolipoprotein E receptor 2 antibody.
Gene ID 396102, 7804, 16975
Concentration 1 mg/ml
Immunogen Synthetic peptide between residues 863-963 of the human ApoER2 protein.
Purification Antigen affinity chromatography
Form Liquid
Storage buffer tris glycine, 0.15M NaCl, 0.05% sodium azide
Storage temperature Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty