|
![]() |
ORDER
SPECIFICATIONS
Antigen symbol | IRE1A |
Antigen name | IRE1 alpha |
Reactivity | Rat, Mouse, Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody type | Primary |
Host | Rabbit |
Isotype | IgG |
Western blot | Yes |
ImmunoFluorescence | Yes |
Antigen synonyms | IRE1a antibody, Inositol-requiring protein 1 antibody, IRE1p antibody, Serine/threonine-protein kinase/endoribonuclease IRE1 antibody, Ire1-alpha anti |
Gene ID | 498013, 78943, 2081 |
Concentration | 1 mg/ml |
Immunogen | Synthetic peptide between amino acids 700-800 of the human IRE1 alpha protein.[Swiss-Prot#075460] |
Purification | Antigen affinity chromatography |
Form | Liquid |
Storage buffer | tris glycine, 0.15M NaCl, 0.05% sodium azide |
Storage temperature | -20° C, Avoid Freeze/Thaw Cycles |
Shipping temperature | Ice |