Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LC3 Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
The reaction with mouse LC3 is weaker than human. The immunogen for this antibody production has 100% sequence homology with rat and zebrafish, 92% with mouse, and 91% with bovine proteins. LC3, a mammalian homologue of Apg8, was originally identified as microtubule-associated protein 1 light chain 3. It is a component of both the MAP1A and MAP1B microtubule-binding domains and the heavy-chain independent regulation of LC3 expression might modify MAP1 microtubule-binding activity during development. However, LC3 is now thought to also be involved in autophagy. LC3-I is cytosolic and LC3-II is membrane bound and enriched in the autophagic vacuole fraction. LC3-II is the first mammalian protein identified that specifically associates with the autophagosome membranes.

Type: Primary
Antigen: LC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Hamster, Primates, Canine, Bacteria, Bovine, Human, Rat, Porcine, Mouse, Zebrafish




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µL 100 µl PIEAPA116930 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol LC3
Antigen name 1A/1B-light chain 3
Reactivity Rat, Human, Dog, Mouse, Zebrafish, Bacteria, Hamster, Bovine, Primates, Porcine
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Antigen synonyms MAP1 light chain 3-like protein 2 a, Microtubule-associated proteins 1A/1B light chain 3B MAP1A/MAP1B LC3 B antibody, MAP1A/b light chain 3 B antibody
Gene ID 81631, 322425, 408001, 67443, 64862, 100462754
Concentration 1 mg/ml
Immunogen Synthetic peptide between amino acids 1-100 of the human LC3 protein.
Purification Antigen affinity chromatography
Form Liquid
Storage buffer PBS, 0.02% sodium azide
Storage temperature -20° C, Avoid Freeze/Thaw Cycles
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping