Anti-SEMA3B Rabbit Polyclonal Antibody

Supplier: Thermo Fisher Scientific
PIEAPA116932EA 0 EUR
PIEAPA116932
Anti-SEMA3B Rabbit Polyclonal Antibody
Antibodies
In Western Blot, a band is seen at ~50 kDa, representing the secreted form of the protein and also a faint band at ~83 kDa, representing the pro-form of the protein. Prior to immunostaining paraffin tissues, antigen retrieval with sodium citrate buffer (pH 6.0) is recommended. Suggested positive control: antigen standard for SEMA3B (transient overexpression lysate), mouse Cerebellum protein, mouse E16 cerebellum lysate. One family of inhibitory axon guidance molecules is the semaphorins. The semaphorins include secreted, transmembrane, and GPI anchored extracellular molecules that are involved in regulating axon guidance by inhibiting axons from growing toward incorrect targets. SEMA 3B inhibits axonal extension by providing local signals to specify territories inaccessible for growing axons.

Type: Primary
Antigen: SEMA3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Canine, Bovine, Human, Mouse, Primates, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR