Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TUB1 Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
Tub-1 protein is C. elegans tubby homolog, required for normal sensory behavior. It is expressed in ciliated neurons and undergoes both cilated and dendritic transport. It has been shown to be involved in regulating fat storage, most likely by modulating transport, sensing, or responding to signals in ciliated neurons. It has also been shown to play a role in regulating life span, but by a different mechanism than fat storage regulation.

Type: Primary
Antigen: TUB1
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG
Reactivity: C. elegans

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µL 100 µl PIEAPA116934 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol TUB1
Reactivity C. elegans
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
Antigen synonyms Tubby protein homolog 1
Gene ID 174312
Format whole serum
Immunogen Recombinant protein corresponding to amino acids 160-421 of Tub-1.
Form Liquid
Storage buffer whole serum, 0.05% sodium azide
Storage temperature -20° C, Avoid Freeze/Thaw Cycles
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty