Anti-POU5F1 Rabbit Polyclonal Antibody

Supplier: Thermo Fisher Scientific
PIEAPA116943EA 0 EUR
PIEAPA116943
Anti-POU5F1 Rabbit Polyclonal Antibody
Antibodies
This antibody is expected to react with swine, feline and bovine samples based on a 93% sequence homology. Stem cells are crucial ingredients in the progressive restriction of cellular developmental capacity during mammalian embryogenesis. Until recently, only embryonic stem cells (ESC) had been shown to exhibit the potential to differentiate into cells of the all three germ layers. However, adult stem cells (ASC) can regain pluripotency (potential to differentiate into various lineages) if transferred from one stem-cell microenvironment into another. The transcription factor OCT4, expressed in ESCs and germ cells, is strongly implicated in the process of maintaining as well as regaining stem-cell pluripotency and functions as a key regulator of mammalian germline development.

Type: Primary
Antigen: POU5F1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Primates, Rat, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR