Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CRYAB Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
This antibody is expected to react with rat, ovine, rabbit, monkey, hamster, bovine and porcine based on sequence homology. Crystallin AB (CryAB) is a developmentally regulated small heat shock protein known to protect cells from stress damage. It may also play an important role in the Golgi reorganization during cell division. CryAB is known to be associated with a number of neurodegenerative pathologies such as Alzheimer disease amd multiple sclerosis. As well, it has been found to be expressed in breast tumors of patients with shorter survival, suggesting that it may be a useful marker in identifying aggressive breast cancer.

Type: Primary
Antigen: CRYAB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µL 100 µl PIEAPA116951 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol CRYAB
Antigen name Crystallin Alpha B
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoFluorescence Yes
Antigen synonyms CryAB antibody, Heat-shock protein beta-5 antibod, Rosenthal fiber componet antibody, Alpha crystallin B chain antibody, Alpha (B)-crystallin antibody
Gene ID 1410
Concentration 0.9 mg/ml
Immunogen Synthetic peptide between amino acids 100-175 of the human Crystallin AB protein.
Purification Antigen affinity chromatography
Form Liquid
Storage buffer tris citrate, 0.1% sodium azide, pH 7-8
Storage temperature 4° C, do not freeze
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping