Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LOX Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
This antibody is 92% homologous to chicken, zebrafish, and xenopus. Lysyl oxidase (LOX), a copper-containing amine oxidase, belongs to a heterogeneous family of enzymes that oxidize primary amine substrates to reactive aldehydes. It plays a vital role in the formation and repair of the extra cellular matrix. As well, LOX is a multifunctional enzyme having diverse biological functions such as developmental regulation, tumor suppression, cell motility and cellular senescence. The secreted form of LOX is responsible for the invasive properties of hypoxic human cancer cells. Thus, it is essential for hypoxia-induced metastasis and is a good therapeutic target for preventing and treating metastases.

Type: Primary
Antigen: LOX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Human, Bovine, Rat, Porcine




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
50 µL 50 µl PIEAPA116955 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol LOX
Antigen name Lysyl oxidase
Reactivity Bovine, Human, Mouse, Porcine, Rat
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
Antigen synonyms Lysyl oxidase antibody., Protein-lysine 6-oxidase antibody, EC 1.4.3.13 antibody
Gene ID 16948, 24914, 100525278, 280841, 4015
Concentration 1 mg/ml
Immunogen Cocktail of two synthetic peptides between amino acids 300-350 and amino acids 200-300 of the human LOX protein.
Purification Affinity chromatography
Form Liquid
Storage buffer PBS, 0.02% sodium azide
Storage temperature -20° C, Avoid Freeze/Thaw Cycles
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty