Anti-AGPAT6 Rabbit Polyclonal Antibody

Supplier: Thermo Fisher Scientific
PIEAPA116961EA 0 EUR
PIEAPA116961
Anti-AGPAT6 Rabbit Polyclonal Antibody
Antibodies
Suggested positive control: mouse skeletal muscle lysate or protein. AGPAT6 is a novel glycerolipid acyltransferase of the ER, which is crucial for the production of milk fat by the mammary gland.

Type: Primary
Antigen: AGPAT6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Primates, Mouse, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR