Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ZMPSTE24 Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
Suggested positive control: human and mouse testis protein or cell lysate. ZMPSTE24 is a zinc metalloprotease involved in the post-translational processing of LMNA precursors. Mutations in ZMPSTE24 have been associated with human diseases, often due to the accumulation of LMNA precursors. It is possible that ZMPSTE24, due to its association with LMNA, plays some role in human ageing.

Type: Primary
Antigen: ZMPSTE24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Zebrafish




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µL 100 µl PIEAPA116964 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol ZMPSTE24
Antigen name Zinc metallopeptidase STE24
Reactivity Zebrafish, Human, Mouse
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoFluorescence Yes
Antigen synonyms EC 3.4.24.84 antibody, Farnesylated proteins-convertin, PyruvaCAAX prenyl protease 1 homolog antibody, Prenyl protein-specific endoprotease 1 antibody
Gene ID 10269, 334380, 230709
Concentration 1 mg/ml
Immunogen Synthetic peptide between amino acids 1-100 of the human ZMPSTE24 protein.
Purification Antigen affinity chromatography
Form Liquid
Storage buffer tris glycine, 150mM NaCl, 0.05% sodium azide
Storage temperature -20° C, Avoid Freeze/Thaw Cycles
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty