Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ITGAL Goat Polyclonal Antibody

Supplier: Thermo Scientific
This antibody is predicted to react with canine based on sequence homology. ITGAL encodes the integrin alpha L chain. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This I-domain containing alpha integrin combines with the beta 2 chain to form the integrin lymphocyte function-associated antigen-1 , which is expressed on all leukocytes. LFA-1 plays a central role in leukocyte intercellular adhesion through interactions with its ligands, ICAMs 1-3 , and also functions in lymphocyte costimulatory signaling. Two transcript variants encoding different isoforms have been found for this gene.

Immunohistochemistry (Paraffin): 4-6µg/ml, Western Blot: 0.5-2µg/ml

Type: Primary
Antigen: ITGAL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µg 100 µG PIEAPA519161 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol ITGAL
Antigen name Integrin, alpha L
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Goat
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
Antigen synonyms integrin alpha-L, antigen CD11A p180, LFA-1 alpha, lymphocyte function-associated antigen 1 alpha polypeptide, LFA-1A, CD11 antigen-like family member A
Gene ID 3683
Concentration 0.5 mg/ml
Immunogen Synthetic peptide sequence (YQVRIQPSIHDHN) corresponding to the internal amino acids of ITGAL
Purification Antigen affinity chromatography
Form Liquid
Storage buffer TBS, 0.5% BSA, 0.02% sodium azide, pH 7.3
Storage temperature -20° C, Avoid Freeze/Thaw Cycles
Shipping temperature Ice
Brand name Invitrogen
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty