Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NUDC Goat Polyclonal Antibody

Supplier: Thermo Scientific
Type: Primary
Antigen: NUDC (nudC nuclear distribution protein)
Clonality: polyclonal
Conjugation: unconjugated
Host: Goat
Isotype: none
Reactivity: Human

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 ug 100 µG PIEAPA519168 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol NUDC
Antigen name Nuclear Distribution Gene C homolog
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Goat
Isotype IgG
Western blot Yes
Antigen synonyms HNUDC, nuclear distribution gene C homolog, OTTHUMP00000004405, nuclear distribution gene C homolog (A. nidulans), MNUDC, NPD011, NUDC
Gene ID 10726
Concentration 0.5 mg/ml
Immunogen Synthetic peptide sequence (PNLGNGADLPNYR) corresponding to the internal amino acids of NUDC
Purification Antigen affinity chromatography
Form Liquid
Storage buffer TBS, 0.5% BSA, 0.02% sodium azide, pH 7.3
Storage temperature -20° C, Avoid Freeze/Thaw Cycles
Shipping temperature Ice
Brand name Invitrogen
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty