Anti-INS Mouse Polyclonal Antibody (Unconjugated) [clone: 2D11.H5]

Supplier: Rockland Immunochemicals
ROCK200-301-389EA 398 EUR
ROCK200-301-389
Anti-INS Mouse Polyclonal Antibody (Unconjugated) [clone: 2D11.H5]
Antibodies
This antibody is suitable for ELISA, immunochemistry, immunoblotting and immunoprecipitation. Insulin has a cytoplasmic localization.  Pancreatic tissue or b-cells from islets of Langerhans can be used as a positive control.

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:50 - 1:200; Immunochemistry: 1:50 - 1:200;

Type: Primary
Antigen: INS
Clonality: Polyclonal
Clone: 2D11.H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR