Anti-StreptAvidin Rabbit Polyclonal Antibody (Peroxidase)

Supplier: Rockland Immunochemicals
ROCK200-4395EA 5560 EUR
ROCK200-4395
Anti-StreptAvidin Rabbit Polyclonal Antibody (Peroxidase)
Antibodies
Type: Tag
Antigen: StreptAvidin
Clonality: Polyclonal
Clone:
Conjugation: Peroxidase
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR