Anti-Avidin Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
ROCK200-4697EA 1960 EUR
ROCK200-4697
Anti-Avidin Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: Avidin
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR