Anti-IgG (H+L) Rabbit Polyclonal Antibody (Peroxidase)

Supplier: Rockland Immunochemicals
ROCK216-4302EA 259 EUR
ROCK216-4302
Anti-IgG (H+L) Rabbit Polyclonal Antibody (Peroxidase)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Donkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR