Anti-IgG F(ab')2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
ROCK312-4104EA 234 EUR
ROCK312-4104
Anti-IgG F(ab')2 Rabbit Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(ab')2
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR