Anti-IgA Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Supplier: Rockland Immunochemicals
ROCK609-4006EA 150 EUR
ROCK609-4006
Anti-IgA Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antibodies
Secondary Rabbit Anti-IgA (alpha chain) Reacts with Human

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgA α
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgA
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR