Anti-IgG (H+L) Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: Rockland Immunochemicals
ROCK609-4502EA 179 EUR
ROCK609-4502
Anti-IgG (H+L) Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Anti-Human IgG antibody is suitable for ELISA,Western Blot and immunochemistry applications. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR