Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rabbit Anti-Human IgA Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
This product has been assayed against 1.0 µg of Human IgA in a standard capture ELISA using Peroxidase Conjugated Streptavidin and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid)) as a substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:250 - 1:1000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgA α
Clonality: Polyclonal
Conjugation: Biotin
Host: Rabbit
Isotype: IgA
Reactivity: Human

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
1 mg 1 mg ROCK609-4606
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol IgA
Antigen name IgA
Reactivity Human
Conjugation Biotin
Clonality Polyclonal
Antibody type Secondary
Host Rabbit
Isotype none
Western blot Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Gel shift Yes
Flow cytometry Yes
ChIP Yes
Neutralisation Yes
Concentration 1,0 mg/ml by UV absorbance at 280 nm
Format Antibody fraction
Cross adsorption non
Immunogen Human IgA alpha heavy chain
Purification purified by immunoaffinity chromatography
Form Lyophilized
Storage buffer 0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Thimerosal, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty