Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sheep Anti-Human IgG (H+L) Polyclonal Antibody (Texas Red®)

Supplier: Rockland Immunochemicals
Secondary Sheep Anti-IgG (H&L) Reacts with Human

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: Heavy and Light chain
Host: Sheep
Isotype: IgG
Reactivity: Human



 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
2 mg 2 mg ROCK609-6902
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol IgG (H+L)
Antigen name IgG (H+L)
Reactivity Human
Conjugation Texas Red®
Clonality Polyclonal
Antibody type Secondary
Host Sheep
Isotype none
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Gel shift Yes
Flow cytometry Yes
ChIP Yes
Neutralisation Yes
Concentration 2,0 mg/ml by UV absorbance at 280 nm
Format Antibody fraction
Cross adsorption non
Immunogen Human IgG whole molecule
Purification purified by immunoaffinity chromatography
Form Lyophilized
Storage buffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty