Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Donkey Anti-Human IgG (H+L) Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: Rockland Immunochemical

This product has been assayed against 1.0 µg of Human IgG in a standard capture ELISA using pNPP (p-nitrophenyl phosphate code # NPP-10) as a substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: Heavy and Light chain
Host: Donkey
Isotype:
Reactivity: Human



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
1 mg 1 mg ROCK609-705-002
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol IgG (H+L)
Antigen name IgG (H+L)
Reactivity Human
Conjugation AP (Alkaline Phosphatase)
Clonality Polyclonal
Antibody type Secondary
Host Donkey
Isotype none
Western Blot Yes
ELISA Yes
ImmunoChemistry Yes
Concentration 1,0 mg/ml by UV absorbance at 280 nm
Format Antibody fraction
Cross Adsorption non
Immunogen Human IgG whole molecule
Purification purified by immunoaffinity chromatography
Form Liquid (sterile filtered)
Storage Buffer 0,05 M Tris Chloride, 0,15M Sodium Chloride, 0,001M Magnesium Chloride, 0,0001M Zinc Chloride, 50% (v/v) Glycerol
Storage temperature Cool
Shipping Temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty