Anti-IgG/IgM (H+L) Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
ROCK610-101-115EA 212 EUR
ROCK610-101-115
Anti-IgG/IgM (H+L) Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:100000; Western Blot: 1:5000 - 1:20000; Immunochemistry: 1:2000 - 1:10000;

Type: Secondary
Antigen: IgG/IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR