Anti-IgM µ Goat Polyclonal Antibody (Texas Red®)

Supplier: Rockland Immunochemicals
ROCK610-1907EA 212 EUR
ROCK610-1907
Anti-IgM µ Goat Polyclonal Antibody (Texas Red®)
Antibodies
Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500; Other Dilution: FLOW CYTOMETRY 1:500 - 1:2500

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: µ-chain
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR