Anti-IgG gamma Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
ROCK610-4112EA 268 EUR
ROCK610-4112
Anti-IgG gamma Rabbit Polyclonal Antibody
Antibodies
Secondary Rabbit Anti-IgG (gamma chain) Reacts with Mouse

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500;

Type: Secondary
Antigen: IgG γ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: γ chain
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR