Anti-IgG gamma Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
ROCK617-106-012EA 218 EUR
ROCK617-106-012
Anti-IgG gamma Goat Polyclonal Antibody (Biotin)
Antibodies
Anti-Monkey IgG (gamma chain) biotin conjugated is suitable for use in immunoelectrophoresis, western-blot, competitive western-blot, ELISA and competitive ELISA assays.

Recommended Dilutions: ELISA: 1:20000 - 1:50000; Western Blot: 1:2000 - 1:5000; Immunochemistry: 1:1000 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG γ
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: γ chain
Host: Goat
Isotype: IgG
Reactivity: Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR