Anti-EPHA5 Mouse Monoclonal Antibody [clone: 13B252]

Supplier: United States Biological
USBI030174EA 506 EUR
USBI030174
Anti-EPHA5 Mouse Monoclonal Antibody [clone: 13B252]
Antibodies
Type: Primary
Antigen: EPHA5 (EPH receptor A5)
Clonality: Monoclonal
Clone: 13B252
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR