Anti-IL6 Mouse Monoclonal Antibody [clone: 13B413]

Supplier: United States Biological
USBI030335EA 506 EUR
USBI030335
Anti-IL6 Mouse Monoclonal Antibody [clone: 13B413]
Antibodies
Type: Primary
Antigen: IL6 (interleukin 6 (interferon, beta 2))
Clonality: Monoclonal
Clone: 13B413
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR