Anti-CACFD1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: US Biological
USBI033089-HRPEA 852 EUR
USBI033089-HRP
Anti-CACFD1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: CACFD1 (calcium channel flower domain containing 1)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR