Anti-CA3 Rabbit Polyclonal Antibody

Supplier: US Biological
USBI033110EA 587 EUR
USBI033110
Anti-CA3 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 50-100; Immunochemistry: 1:50-100

Type: Primary
Antigen: CA3 (carbonic anhydrase III, muscle specific)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR