Anti-CACNA2D2 Rabbit Polyclonal Antibody

Supplier: US Biological
USBI033127EA 587 EUR
USBI033127
Anti-CACNA2D2 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Immunofluorescence: 1:10-1:50; Western Blot: 1:1000; Immunochemistry: 1:10-1:50

Type: Primary
Antigen: CACNA2D2 (calcium channel, voltage-dependent, alpha 2/delta subunit 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR