Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MMP14 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: US Biological
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: MMP14 (matrix metallopeptidase 14 (membrane-inserted))
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
200 µl 200 µl USBI038501-HRP
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol MMP14
Antigen name matrix metallopeptidase 14 (membrane-inserted)
Reactivity Human
Conjugation HRP (Horseradish Peroxidase)
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Amino acid number 495 to 523
Epitope C-terminal
Purification purified by Protein A chromatography
Form liquid
Storage buffer PBS, pH 7,2
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty