Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MMP15 Rabbit Polyclonal Antibody (Biotin)

Supplier: US Biological
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50

Type: Primary
Antigen: MMP15 (matrix metallopeptidase 15 (membrane-inserted))
Clonality: Polyclonal
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
200 µl 200 µl USBI038503-BIOTIN
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol MMP15
Antigen name matrix metallopeptidase 15 (membrane-inserted)
Reactivity Human
Conjugation Biotin
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
Amino acid number 167 to 196
Epitope N-terminal
Purification purified by Protein A chromatography
Form liquid
Storage buffer PBS, pH 7,2
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty