Anti-NAA50 Rabbit Polyclonal Antibody (Biotin)

Supplier: US Biological
038805-BIOTIN
USBI038805-BIOTINEA 852 EUR
USBI038805-BIOTIN
Anti-NAA50 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50

Type: Primary
Antigen: NAA50 (N(alpha)-acetyltransferase 50, NatE catalytic subunit)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR