Anti-NCAM2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Supplier: US Biological
038820-APC
USBI038820-APCEA 852 EUR
USBI038820-APC
Anti-NCAM2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: NCAM2 (neural cell adhesion molecule 2)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR