Anti-MT-ND3 Rabbit Polyclonal Antibody

Supplier: US Biological
038840
USBI038840EA 587 EUR
USBI038840
Anti-MT-ND3 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50

Type: Primary
Antigen: MT-ND3 (mitochondrially encoded NADH dehydrogenase 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR