Anti-NDUFS2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Supplier: US Biological
038869-APC
USBI038869-APCEA 852 EUR
USBI038869-APC
Anti-NDUFS2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!

Type: Primary
Antigen: NDUFS2 (NADH dehydrogenase (ubiquinone) Fe-S protein 2, 49kDa (NADH-coenzyme Q reductase))
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR