Anti-NEK6 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: US Biological
038888-FITC
USBI038888-FITCEA 852 EUR
USBI038888-FITC
Anti-NEK6 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: NEK6 (NIMA-related kinase 6)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR