Anti-NRP2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Supplier: US Biological
038920-PE
USBI038920-PEEA 852 EUR
USBI038920-PE
Anti-NRP2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: NRP2 (neuropilin 2)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR