Anti-NHEJ1 Rabbit Polyclonal Antibody

Supplier: US Biological
USBI038960EA 587 EUR
USBI038960
Anti-NHEJ1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: NHEJ1 (nonhomologous end-joining factor 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR