Anti-NKIRAS1 Rabbit Polyclonal Antibody (Biotin)

Supplier: US Biological
038990-BIOTIN
USBI038990-BIOTINEA 852 EUR
USBI038990-BIOTIN
Anti-NKIRAS1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: NKIRAS1 (NFKB inhibitor interacting Ras-like 1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR