Anti-NKPD1 Rabbit Polyclonal Antibody (Biotin)

Supplier: US Biological
038992-BIOTIN
USBI038992-BIOTINEA 852 EUR
USBI038992-BIOTIN
Anti-NKPD1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50

Type: Primary
Antigen: NKPD1 (NTPase, KAP family P-loop domain containing 1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR