Anti-NKX6-2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Supplier: US Biological
USBI039008-PEEA 852 EUR
USBI039008-PE
Anti-NKX6-2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: NKX6-2 (NK6 homeobox 2)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR