Anti-NOC3L Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Supplier: US Biological
039040-APC
USBI039040-APCEA 852 EUR
USBI039040-APC
Anti-NOC3L Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: NOC3L (nucleolar complex associated 3 homolog (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR