Anti-NOTCH1 Rabbit Polyclonal Antibody (Biotin)

Supplier: US Biological
039065-BIOTIN
USBI039065-BIOTINEA 852 EUR
USBI039065-BIOTIN
Anti-NOTCH1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:50-100

Type: Primary
Antigen: NOTCH1 (notch 1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR