Anti-NRG3 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Supplier: US Biological
039148-PE
USBI039148-PEEA 852 EUR
USBI039148-PE
Anti-NRG3 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Recommended dilution: FLISA: 1:1000; Immunochemistry: 1:50-100
Do not freeze!

Type: Primary
Antigen: NRG3 (neuregulin 3)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR