Anti-NTRK3 Rabbit Polyclonal Antibody (Biotin)

Supplier: US Biological
039186-BIOTIN
USBI039186-BIOTINEA 852 EUR
USBI039186-BIOTIN
Anti-NTRK3 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50

Type: Primary
Antigen: NTRK3 (neurotrophic tyrosine kinase, receptor, type 3)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR